Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant CCND1 (from HEK293 cells)
Catalog # 77538-850
Supplier:  Creative Biomart
New Product
CAS Number:  
undefined
Human Recombinant CCND1 (from HEK293 cells)
Catalog # 77538-850
Supplier:  Creative Biomart
Supplier Number:  CCND1-001HB
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    10 µg
  • Storage Conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-Free:
    No
  • Protease-free:
    No
  • Animal-Free:
    No
  • Protein Synonyms:
    cyclin D1
  • UniProtKB:
    P24385
  • Protein/Peptide Name:
    CCND1
  • Purity:
    >90% by SDS-PAGE
  • Molecular Weight:
    The protein has a calculated MW of 37 kDa
  • Sequence:
    MEHQLLCCEVETIRRAYPDANLLNDRVLRAMLKAEETCAPSVSYFKCVQKEVLPSMRKIVATWMLEVCEEQKCEEEVFPLAMNYLDRFLSLEPVKKSRLQLLGATCMFVASKMKETIPLTAEKLCIYTDNSIRPEELLQMELLLVNKLKWNLAAMTPHDFIEHFLSKMPEAEENKQIIRKHAQTFVALCATDVKFISNPPSMVAAGSVVAAVQGLNLRSPNNFLSYYRLTRFLSRVIKCDPDCLRACQEQIEALLESSLRQAQQNMDPKAAEEEEEEEEEVDLACTPTDVRDVDIHHHHHHHHGLNDIFEAQKIEWHE
  • Endotoxin Level:
    Low
  • Concentration:
    1.39 mg/ml
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping Temperature:
    20
  • Cat. No.:
    77538-850

Specifications

About this item