Order Entry
United States
Orders LinkContactUsLinkComponent
 

Human Native BGLAP (from Plasma)

Vial, 20 µg
Catalog # 77538-736
Supplier:   Creative Biomart
 :  BGLAP-60H
Human Native BGLAP (from Plasma)
Catalog # 77538-736
Supplier:  Creative Biomart
CAS Number:  
Human Native BGLAP (from Plasma)
Human Native BGLAP (from Plasma)

Vial, 20 µg

CAS Number:  
Price
$424.29
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    20 µg
  • Storage Conditions:
    4 °C
  • Molecular Weight:
    5800
  • Protease-free:
    No
  • Endotoxin Level:
    Low
  • Animal-Free:
    No
  • UniProtKB:
    P02818
  • Purity:
    >95% by SDS-PAGE
  • Carrier-Free:
    No
  • Grade:
    Ultrapure grade
  • Concentration:
    0.68 mg/ml
  • Source:
    Plasma
  • Nuclease-free:
    No
  • Cat. No.:
    77538-736
  • Shipping Temperature:
    20
  • Protein/Peptide Type:
    Native
  • Protein Synonyms:
    bone gamma-carboxyglutamate (gla) protein
  • Protein/Peptide Name:
    BGLAP
  • Conjugation:
    Unconjugated
  • Endotoxin-free:
    No
  • Species:
    Human

Specifications