Human Recombinant ASS1 (from E. coli)
Catalog # 77538-686
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:10 MU
- Storage Conditions:4 °C
- Biological Activity:The specific activity is 500 mU/mg based on its ability to convert citrulline to argininosuccinate
- Reconstitution Instructions:Reconstitute the lyophilized protein in H₂O with concentration of 1 mU/ml. Incubate the reconstituted protein at 25 centigrade for 15 minutes.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:argininosuccinate synthase 1
- UniProtKB:P00966
- Protein/Peptide Name:ASS1
- Purity:≥95% (SDS-PAGE)
- Molecular Weight:50 kDa including tags
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Tested Applications:SDS-PAGE, functional studies
- Cat. No.:77538-686