Human Recombinant AHR (from E. coli)
Catalog # 77538-576
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:10 µg
- Storage Conditions:4 °C
- Endotoxin Content:<1 EU/1 µg (determined by the LAL method)
- Reconstitution Instructions:Reconstitute in PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:aryl hydrocarbon receptor
- UniProtKB:P35869
- Protein/Peptide Name:AHR
- Purity:>97%
- Molecular Weight:32.2 kDa
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77538-576