Order Entry
United States
ContactUsLinkComponent
 
Halogen Free Ribbons for M6 Printer, R6000
  77574-914
New Product
undefined
Halogen Free Ribbons for M6 Printer, R6000
  77574-914
 :  176977
New Product

 

  • For Use With:
    M6 Printers
  • Color:
    Black
  • Description:
    Halogen free ribbon, R6000 series
  • Length:
    22.86 m
  • Cat. No.:
    77574-914
  • Width:
    50.80 mm

 

 

M6 Series ribbon cartridges allow users to change ribbons in seconds.

  • One-color ribbon for printing a single color onto a label
  • R6300 Series Halogen-Free Resin Formulation offers excellent solvent, scratch and heat resistance when printed on the appropriate label material
  • Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra Aggressive Polyester and B-423 Permanent Polyester from Brady®

Automatic label formatting with smart cell technology and easy loading.