Gelite™ Safe DNA Gel Stain *GelRed Alternative, 10000X in Water*
77461-666
:
- Description:Gelite™ safe DNA gel stain *GelRed alternative, 10000X in water*
- Size:0.5 ml
- Cat. No.:77461-666
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity.
- Stable for 12 months
AAT Bioquest is committed to designing our products to be environment-friendly.
: For research use only.