ContactUsLinkComponent
Gelite™ Safe DNA Gel Stain *10000X Water Solution*
Gelite™ Safe DNA Gel Stain *10000X Water Solution*
Catalog # 77461-660
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
Gelite™ Safe DNA Gel Stain *10000X Water Solution*
Catalog # 77461-660
Supplier:  AAT BIOQUEST INC
Supplier Number:  17699
CAS Number:  
Specifications
  • Description:
    Gelite™ safe DNA gel stain *10000X water solution*
  • Size:
    50 µl
  • Cat. No.:
    77461-660
Specifications
About this item
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity.
  • Stable for 12 months
AAT Bioquest is committed to designing our products to be environment-friendly.
Caution: For research use only.