ContactUsLinkComponent
Gelite™ 100 bp to 1 kb DNA Ladder
Gelite™ 100 bp to 1 kb DNA Ladder
  77461-676
 :  
undefined
Gelite™ 100 bp to 1 kb DNA Ladder
  77461-676
 :  17760
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Gelite™ 100 bp - 1 kb DNA ladder
  • Size:
    0.5 ml
  • Environmentally Preferable:
  • Cat. No.:
    77461-676
The Gelite™ 100 bp to 1 kb DNA ladder is designed to help size and estimate the mass of double-stranded DNA and PCR products within the 100 to 10000 base pair range on agarose or polyacrylamide gels.
  • Stable for 12 months
Quality tested in-house.
 : For research use only.