Order Entry
United States
ContactUsLinkComponent
Universal Rotary Circuit Breaker Lockout
Universal Rotary Circuit Breaker Lockout
Catalog # 77574-856
Supplier:  Brady Worldwide
New Product
undefined
Universal Rotary Circuit Breaker Lockout
Catalog # 77574-856
Supplier:  Brady Worldwide
Supplier Number:  177243
New Product

Specifications

  • Color:
    Red, Black
  • Description:
    Universal rotary circuit breaker lockout
  • Dimensions:
    38.10W×38.10D×88.9H mm
  • Material:
    Polypropylene
  • Cat. No.:
    77574-856

Specifications

About this item

Enhance worker safety and maximise lockout efficiency with the Universal Rotary Breaker Lockout – the most adaptable rotary switch device for streamlined safety and compliance.

  • Universal fit securely accommodates most engine switches with a panel height up to 45 mm
  • Intuitive thumbwheel design ensures fast, tool-free installation and removal
  • Industrial-grade construction withstands harsh environments
  • Meets requirements as part of an OSHA-compliant lockout tagout program