Human Recombinant LAMA5 (from E. coli), Biotinlyted
Catalog # 77540-098
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:laminin, alpha 5
- UniProtKB:O15230
- Protein/Peptide Name:LAMA5
- Endotoxin Level:Low
- Concentration:250 µg/2.75 ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Tested Applications:Cell culture of murine embryonic and induced pluripotent stem cells
- Cat. No.:77540-098