Order Entry
United States
ContactUsLinkComponent
Human Recombinant IL18 (from HEK293 cells)
Human Recombinant IL18 (from HEK293 cells)
Supplier:  Creative Biomart
New Product
Certificates
undefined
Human Recombinant IL18 (from HEK293 cells)
Supplier:  Creative Biomart
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    4 °C
  • Endotoxin Content:
    <100 EU/mg of the protein as determined by the LAL method
  • Biological Activity:
    1.0×10⁶ IU, Measured by its ability to induce IFN-gamma secretion by KG‐1 human acute myelogenous leukemia cells in the presence of TNF-alpha
  • Reconstitution Instructions:
    Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration >100μg/ml. Dissolve the lyophilized protein in deionised H₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Endotoxin-free:
    No
  • Carrier-Free:
    No
  • Protease-free:
    No
  • Animal-Free:
    No
  • Protein Synonyms:
    interleukin 18
  • UniProtKB:
    Q14116
  • Protein/Peptide Name:
    IL18
  • Purity:
    >95% as determined by SDS-PAGE
  • Molecular Weight:
    The apparent molecular mass of recombinant Human IL18 is approximately 22 kDa in SDS-PAGE under reducing conditions
  • Sequence:
    YFGKLESKLS VIRNLNDQVL FIDQGNRPLF EDMTDSDCRD NAPRTIFIISMYKDSQPRGM AVTISVKCEK ISTLSCENKI ISFKEMNPPD NIKDTKSDIIFFQRSVPGHD NKMQFESSSY EGYFLACEKE RDLFKLILKK EDELGDRSIMFTVQNEDVD
  • Endotoxin Level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping Temperature:
    20

Specifications

Frequently Bought Together