Human Recombinant IDH1 (from E. coli)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:isocitrate dehydrogenase 1 (NADP+), soluble
- UniProtKB:O75874
- Protein/Peptide Name:IDH1
- Purity:≥95% as determined by SDS-PAGE
- Molecular Weight:47.9 kDa
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
Specifications
Frequently Bought Together