Mouse Recombinant Hmgb1 (from HEK293 cells)
Catalog # 77539-772
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:10 µg
- Storage Conditions:4 °C
- Endotoxin Content:<1.0 EU/µg of the protein as determined by the LAL method
- Biological Activity:Measured by its ability to bind mouse AGER-His in functional ELISA
- Reconstitution Instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:high mobility group box 1
- Protein/Peptide Name:Hmgb1
- Purity:>85% as determined by SDS-PAGE
- Molecular Weight:The recombinant mouse HMGB1/Fc is a disulfide-linked homodimer. The reduced monomer comprises 475 amino acids and has a calculated molecular mass of 53.3 kDa. The apparent molecular mass of the monomer is approximately 57 kDa in SDS-PAGE under reducing conditions.
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-772