Human Native HeLa (from Epidermoid carcinoma)
Catalog # 77539-734
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Native
- Source:Epidermoid carcinoma
- Species:Human
- Size:200 µg
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:HeLa Cell Nuclear Extract
- Protein/Peptide Name:HeLa
- Endotoxin Level:Low
- Concentration:2 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Tested Applications:Protein lysate for WB. Ready-to-use lysates are especially prepared as positive controls for separation by SDS-PAGE and subsequent western blot analysis. Lysates are prepared in denaturing buffer WITHOUT dissociating agents (i.e. no 2-mercaptoethanol or dithiothreitol has been added).
- Cat. No.:77539-734