Human Recombinant GPX4 (from E. coli)
Supplier: Creative Biomart
New Product
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:4 °C
- Endotoxin Content:<1 EU/µg
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:glutathione peroxidase 4
- UniProtKB:P36969
- Protein/Peptide Name:GPX4
- Purity:>90%, by SDS-PAGE
- Molecular Weight:~21.3 kDa, reducing conditions
- Sequence:MHHHHHHENLYFQSMCASRDDWRCARSMHEFSAKDIDGHMVNLDKYRGFVCIVTNVASQCGKTEVNYTQLVDLHARYAECGLRILAFPCNQFGKQEPGSNEEIKEFAAGYNVKFDMFSKICVNGDDAHPLWKWMKIQPKGKGILGNAIKWNFTKFLIDKNGCVVKRYGPMEEPLVIEKDLPHYF
- Endotoxin Level:Low
- Concentration:0.125 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
Specifications
Frequently Bought Together