Order Entry
United States
ContactUsLinkComponent
HTLV-1 Recombinant GB46 (from HEK293 cells)
HTLV-1 Recombinant GB46 (from HEK293 cells)
Supplier:  Creative Biomart
Certificates
undefined
HTLV-1 Recombinant GB46 (from HEK293 cells)
Supplier:  Creative Biomart

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    HTLV-1
  • Storage Conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-Free:
    No
  • Protease-free:
    No
  • Animal-Free:
    No
  • Protein Synonyms:
    gp46 protein
  • Protein/Peptide Name:
    GB46
  • Purity:
    >90% by SDS-PAGE
  • Molecular Weight:
    The protein has a calculated MW of 32.2 kDa
  • Sequence:
    SCCTLTIGVSSYHSKPCNPAQPVCSWTLDLLALSADQALQPPCPNLVSYSSYHATYSLYLFPHWTKKPNRNGGGYYSASYSDPCSLKCPYLGCQSWTCPYTGAVSSPYWKFQHDVNFTQEVSRLNINLHFSKCGFPFSLLVDAPGYDPIWFLNTEPSQLPPTAPPLLPHSNLDHILEPSIPWKSKLLTLVQLTLQSTNYTCIVCIDRASLSTWHVLYSPNVSVPSSSSTPLLYPSLALPAPHLTLPFNWTHCFDPQIQAIVSSPCHNSLILPPFSLSPVPTLGSHHHHHH
  • Endotoxin Level:
    Low
  • Concentration:
    0.63 mg/ml
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping Temperature:
    20

Specifications

Frequently Bought Together