Human Recombinant FLT3 (from HEK293 cells)
Catalog # 77539-524
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:50 µg
- Storage Conditions:4 °C
- Endotoxin Content:<1.0 EU/µg of the protein as determined by the LAL method
- Biological Activity:Measured by its ability to bind biotinylated human FLT3L-His in a functional ELISA
- Reconstitution Instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:fms-related tyrosine kinase 3
- UniProtKB:P36888
- Protein/Peptide Name:FLT3
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The recombinant human FLT3 consists of 526 amino acids and has a predicted molecular mass of 59.7 kDa. In SDS-PAGE under reducing conditions, it migrates with an apparent molecular mass of 85 - 95 kDa due to glycosylation.
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-524