Human Recombinant FCGR1A (from HEK293 cells)
77539-450
New Product
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:50 µg
- Storage Conditions:4 °C
- Endotoxin Content:<1.0 EU/µg of the protein as determined by the LAL method
- Biological Activity:Immobilized Rituximab at 1 μg/ml can bind Human FcγRI/CD64 recombinant protein, with a linear range of 0.5 - 4.0 ng/ml. Captured Human FcγRI/CD64 recombinant protein on Anti-His Chip can bind Bevacizumab (IgG1) with an affinity constant of 5.5 nM as determined in an SPR assay (Biacore T200). Captured Human FcγRI/CD64 recombinant protein on Anti-His Chip can bind Nivolumab (IgG4) with an affinity constant of 13.6 nM as determined in an SPR assay (Biacore T200). Loaded Human FcγRI/CD64 recombinant protein on His1K Biosensor, can bind Nivolumab (IgG4) with an affinity constant of 8.5 nM as determined in a BLI assay (ForteBio Octet Red384). Loaded Human FcγRI/CD64 recombinant protein on His1K biosensor, can bind Bevacizumab (IgG1) with an affinity constant of 4.7 nM as determined in a BLI assay (ForteBio Octet Red384). Loaded Rituximab (IgG1) on ProA biosensor, can bind Human FcγRI/CD64 recombinant protein with an affinity constant of 0.38 nM as determined in a BLI assay (ForteBio Octet Red384).
- Reconstitution Instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:Fc fragment of IgG, high affinity Ia, receptor (CD64)
- UniProtKB:P12314
- Protein/Peptide Name:FCGR1A
- Purity:>95% as determined by SDS-PAGE. >90% as determined by SEC-HPLC.
- Molecular Weight:The recombinant human CD64A consists of 284 amino acids and has a predicted molecular mass of 32 kDa. In SDS-PAGE under reducing conditions, the apparent molecular mass of the recombinant human CD64A is approximately 45 - 60 kDa due to glycosylation.
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-450