Human Recombinant EIF2AK2 (from Insect cells)
Catalog # 77539-350
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Size:5 µg
- Storage Conditions:4 °C
- Biological Activity:The activity was determined by IMAPTM assay. The enzyme was incubated with fluorescein labeled peptide and phosphorylation was detected by IMAPTM technology (fluorescence polarization). Substrate: SRPKtide. ATP: 100 μM.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:eukaryotic translation initiation factor 2-alpha kinase 2
- UniProtKB:P19525
- Protein/Peptide Name:EIF2AK2
- Purity:61% by SDS-PAGE/CBB staining
- Molecular Weight:62 kDa
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Tested Applications:Functional study, SDS-PAGE
- Cat. No.:77539-350