Order Entry
United States
ContactUsLinkComponent
Human Recombinant DPM1 (from E. coli)
Human Recombinant DPM1 (from E. coli)
Supplier:  Creative Biomart
New Product
Certificates
undefined
Human Recombinant DPM1 (from E. coli)
Supplier:  Creative Biomart
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-Free:
    No
  • Protease-free:
    No
  • Animal-Free:
    No
  • Protein Synonyms:
    dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit
  • UniProtKB:
    O60762
  • Protein/Peptide Name:
    DPM1
  • Purity:
    >90% by SDS-PAGE
  • Molecular Weight:
    The protein has a calculated MW of 30.45 kDa
  • Sequence:
    HHHHHHMASLEVSRSPRRSR RELEVRSPRQ NKYSVLLPTYNERENLPLIV WLLVKSFSESGINYEIIIID DGSPDGTRDVAEQLEKIYGSDRILLRPREK KLGLGTAYIHGMKHATGNYIIIMDADLSHHPKFIPEFIRKQKEGNFDIVS GTRYKGNGGVYGWDLKRKIISRGANFLTQILLRPGASDLTGSFRLYRKEV LEKLIEKCVSKGYVFQMEMIVRARQLNYTIGEVPISFVDRVYGESKLGGN EIVSFLKGLLTLFATT
  • Endotoxin Level:
    Low
  • Concentration:
    0.4 mg/ml
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping Temperature:
    20

Specifications

Frequently Bought Together