Human Recombinant DNA2 (from E. coli)
Catalog # 77539-302
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:25 µg
- Storage Conditions:4 °C
- Reconstitution Instructions:Reconstitute at 0.25 µg/μl in 200 μl sterile water for short-term storage.Reconstitution with 200 μl 50% glycerol solution is recommended for longer term storage.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:DNA replication helicase/nuclease 2
- UniProtKB:P51530
- Protein/Peptide Name:DNA2
- Purity:85% by SDS-PAGE with Coomassie brilliant blue staining
- Sequence:MEQLNELELLMEKSFWEEAELPAELFQKKVVASFPRTVLSTGMDNRYLVLAVNTVQNKEGNCEKRLVITASQSLENKELCILRNDWCSVPVEPGDIIHLEGDCTSDTWIIDKDFGYLILYPDMLISGTSIASSIRCMRRAVLSETFRSSDPATRQMLIGTVLHEVFQKAINNSFAPEKLQELAFQTIQEIRHLKEMYRLNLSQDEIKQEVEDYLPSFCKWAGDFMHKNTSTDFPQMQLSLPSDNSKDNSTCNIEVVKPMDIEESIWSPRFGLKGKIDVTVGVKIHRGYKTKYKIMPLELKTGKESNSIEHRSQVVLYTLLSQERRADPEAGLLLYLKTGQMYPVPANHLD
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-302