Human Recombinant CDK1 (from Insect cells)
Catalog # 77539-016
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Size:50 µg
- Storage Conditions:4 °C
- Endotoxin Content:<1.0 EU/µg protein as determined by the LAL method
- Reconstitution Instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:cyclin-dependent kinase 1
- UniProtKB:P06493
- Protein/Peptide Name:CDK1
- Purity:>85% as determined by SDS-PAGE
- Molecular Weight:The recombinant human CDC2/GST chimera consists of 522 amino acids and predicts a molecular mass of 60 kDa. It migrates as an approximately 53 kDa band in SDS-PAGE under reducing conditions.
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-016