Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant CD33 (from Mammalian cells), FITC (Fluorescein Isothiocyanate)
 
undefined
Human Recombinant CD33 (from Mammalian cells), FITC (Fluorescein Isothiocyanate)

 

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Mammalian cells
  • Species:
    Human
  • Storage Conditions:
    4 °C
  • Biological Activity:
    Measured by the ability of the immobilized protein to support the adhesion of human red blood cells. The ED50 for this effect is typically 1 - 4 μg/ml.
  • Endotoxin-free:
    No
  • Carrier-Free:
    No
  • Protease-free:
    No
  • Animal-Free:
    No
  • Protein Synonyms:
    CD33 molecule
  • UniProtKB:
    P20138
  • Protein/Peptide Name:
    CD33
  • Purity:
    >90% by SDS-PAGE and analyzed by silver stain
  • Molecular Weight:
    Recombinant Human CD33, Fc Chimera has a calculated MW of 53.4 kDa (monomer). In SDS-PAGE migrates as 67 - 85 kDa, reducing conditions.
  • Endotoxin Level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping Temperature:
    20

 

Frequently Bought Together