Pig Recombinant ANGPT2 (from Mammalian cells)
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:Pig
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:angiopoietin 2
- UniProtKB:Q9BDY7
- Protein/Peptide Name:ANGPT2
- Purity:>90% by SDS-PAGE
- Molecular Weight:56 kDa
- Sequence:HHHHHHHHYNNFRKSMDSTGKRQYQVQHGPCSYTFLLPETDNCRSPSSSYVSNAVQRDAPLDYDDSVRRLQVLENIMENNTQWLMKLESYIQDNMKKEMVEIQQNAVQNQTAVMIEIGTNLLNQTAEQTRKLTDVEAQVLNQTTRLELQLLEHSLSTNKLEKQILDQTSEINKLQDKNSFLEKKVLDMEDKHIVQLQSIKEEKDQLQVLVSKQNSIIEELEKQLVTATVNNSVLQKQQHDLMETVHNLLTMISTSNSAKHSLVAKEEQIIFRDCAEAFKSGLTTSGTYTLTFPNSTEETKAYCDMETGGGGWTVIQRREDGSVDFQRTWKEYKMGFGSPSGEHWLGNEFVSQVTNQKRYVLKIHLRDWEGNEAYSLYEHFYLSSEEFNYRIHLKGLTGTAGKISSISQPGNDFSTKDADNDKCICKCSQMLTGGWWFDACGPSNLNGMYYPQRQNTNKFNGIKWYYWKGSGYSLKATTMMIRPADF
- Endotoxin Level:Low
- Concentration:0.19 mg/ml by BCA
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
Frequently Bought Together