Human Recombinant ALKBH3
Catalog # 77538-598
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:10 µg
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:alkB, alkylation repair homolog 3 (E. coli)
- UniProtKB:Q96Q83
- Protein/Peptide Name:ALKBH3
- Purity:>95% by SDS-PAGE
- Endotoxin Level:Low
- Concentration:1 mg/ml (determined by Bradford assay)
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77538-598