Recombinant Human INHBC (from HEK293 cells)
77540-014
New Product
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:25 µg
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:inhibin, beta C
- UniProtKB:P55103
- Protein/Peptide Name:INHBC
- Purity:>90%, by SDS-PAGE
- Molecular Weight:The protein has a calculated MW of 14.2 kDa.
- Sequence:HHHHHHHHDDDDKGIDCQGGSRMCCRQEFFVDFREIGWHDWIIQPEGYAMNFCIGQCPLHIAGMPGIAASFHTAVLNLLKANTAAGTTGGGSCCVPTARRPLSLLYYDRDSNIVKTDIPDMVVEACGCS
- Endotoxin Level:Low
- Concentration:0.27 mg/ml by BCA
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77540-014