Recombinant Human IL7 (from Human Cells)
Supplier: Creative Biomart
New Product
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Human cells
- Species:Human
- Storage Conditions:4 °C
- Endotoxin Content:<0.1 EU/μg
- Biological Activity:2.0×10⁶ U/mg
- Reconstitution Instructions:Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml.Dissolve the lyophilized protein in 3X PBS. Please aliquot the reconstituted solution.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:interleukin 7
- UniProtKB:P13232
- Protein/Peptide Name:IL7
- Purity:>95% as determined by SEC-HPLC and reducing SDS-PAGE
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
Specifications
Frequently Bought Together