Order Entry
United States
ContactUsLinkComponent
Recombinant Human IL1B (from E. coli) R-PE
Recombinant Human IL1B (from E. coli) R-PE
Catalog # 77539-930
Supplier:  Creative Biomart
New Product
CAS Number:  
undefined
Recombinant Human IL1B (from E. coli) R-PE
Catalog # 77539-930
Supplier:  Creative Biomart
Supplier Number:  IL1B-02HP
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    RPE (R-Phycoerythrin)
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    10 µg
  • Storage Conditions:
    4 °C
  • Endotoxin Content:
    <1 EU/µg of rHuIL-1β as determined by LAL method
  • Biological Activity:
    Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells. The ED50 for this effect is less or equal 12 pg/ml.
  • Endotoxin-free:
    No
  • Carrier-Free:
    No
  • Protease-free:
    No
  • Animal-Free:
    No
  • Protein Synonyms:
    IL1 beta
  • UniProtKB:
    P01584
  • Protein/Peptide Name:
    IL1B
  • Purity:
    >95% by SDS-PAGE
  • Molecular Weight:
    Approximately 17.3 kDa, a single non-glycosylated polypeptide chain containing 154 amino acids.
  • Sequence:
    MAPVRSLNCTL RDSQQKSLVM SGPYELKALH LQGQDMEQQV VFSMSFVQGE ESNDKIPVAL GLKEKNLYLS CVLKDDKPTL QLESVDPKNY PKKKMEKRFV FNKIEINNKL EFESAQFPNW YISTSQAENM PVFLGGTKGG QDITDFTMQF VSS
  • Endotoxin Level:
    Low
  • Concentration:
    1.0 mg/ml
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping Temperature:
    20
  • Cat. No.:
    77539-930

Specifications

About this item