Recombinant Human IGHMBP2 (from E. coli)
Catalog # 77539-864
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:25 µg
- Storage Conditions:4 °C
- Endotoxin Content:<1 EU/μg by LAL
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:immunoglobulin mu binding protein 2
- UniProtKB:P38935
- Protein/Peptide Name:IGHMBP2
- Purity:>90%, by SDS-PAGE
- Molecular Weight:The protein has a calculated MW of 110 kDa.
- Endotoxin Level:Low
- Concentration:0.2 mg/ml by BCA
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-864