Recombinant Bovine Ifna (from Yeast)
Catalog # 77539-848
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Yeast
- Species:Bovine
- Size:25 µg
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:glycoprotein hormones, alpha polypeptide/chorionic gonadotropin, beta polypeptide
- UniProtKB:P05007
- Protein/Peptide Name:Ifna
- Purity:>95% as visualized by SDS-PAGE analysis
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-848