Recombinant Human IFNA2 (from E. coli)
Supplier: Creative Biomart
New Product
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:4 °C
- Endotoxin Content:<0.1 ng/µg (1 IEU/µg) of PEG-IFN alpha-2a
- Reconstitution Instructions:It is recommended to reconstitute the lyophilized PEG-IFN alpha-2a in sterile 18M.-cm H₂O not less than 100 µg/ml, which can then be further diluted to other aqueous solutions.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:IFNA2 interferon, alpha 2
- UniProtKB:P01563
- Protein/Peptide Name:IFNA2
- Purity:>98% as determined by HPLC, FPLC and SDS-PAGE analysis
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
Specifications
Frequently Bought Together