Recombinant Human HAVCR1 (from CHO cells)
77539-682
New Product
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Size:10 µg
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:hepatitis A virus cellular receptor 1
- UniProtKB:Q96D42
- Protein/Peptide Name:HAVCR1
- Purity:≥98% (SDS-PAGE)
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-682