Recombinant E.coli HAAO (from Human)
Supplier: Creative Biomart
New Product
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Human cells
- Species:E. coli
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:3-hydroxyanthranilate 3,4-dioxygenase
- UniProtKB:P46952
- Protein/Peptide Name:HAAO
- Purity:>90%, by SDS-PAGE
- Endotoxin Level:Low
- Concentration:0.5 mg/ml (determinedby BCA assay)
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
Specifications
Frequently Bought Together