Recombinant Human GFP (from E. coli)
77539-594
New Product
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:25 µg
- Environmentally Preferable:
- Storage Conditions:4 °C
- Endotoxin Content:<1.0 EU/μg (determined by the LAL method)
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- UniProtKB:P42212
- Protein/Peptide Name:GFP
- Purity:>95% by SDS-PAGE
- Sequence:KETWWETWWTEWSQPKKKRKGMSKGEELFTGVVPILVELDGDVNGHKFSVSGEGEGDATYGKLTLKFICTTGKLPVPWPTLVTTFSYGVQCFSRYPDHMKQHDFFKSAMPEGYVQERTIFFKDDGNYKTRAEVKFEGDTLVNRIELKGIDFKEDGNILGHKLEYNYNSHNVYIMADKQKNGIKVNFKIRHNIEDGSVQLADHYQQNTPIGDGPVLLPDNHYLSTQSALSKDPNEKRDHMVLLEFVTAAGITHGMDELYKTRTRPLEQKLISEEDLAANDIL DYKDDDDKV
- Endotoxin Level:Low
- Concentration:0.1 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-594