Order Entry
United States
ContactUsLinkComponent
Recombinant Human FZD6 (from HEK293 cells)
Recombinant Human FZD6 (from HEK293 cells)
Catalog # 77539-552
Supplier:  Creative Biomart
New Product
CAS Number:  
undefined
Recombinant Human FZD6 (from HEK293 cells)
Catalog # 77539-552
Supplier:  Creative Biomart
Supplier Number:  FZD6-739HB
New Product
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    100 µg
  • Storage Conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-Free:
    No
  • Protease-free:
    No
  • Animal-Free:
    No
  • Protein Synonyms:
    frizzled class receptor 6
  • Protein/Peptide Name:
    FZD6
  • Endotoxin Level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping Temperature:
    20
  • Cat. No.:
    77539-552

Specifications

About this item