Recombinant Human FSHB (from Wheat Germ)
Catalog # 77539-544
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Wheat germ
- Species:Human
- Size:2 µg
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:follicle stimulating hormone, beta polypeptide
- UniProtKB:P01225
- Protein/Peptide Name:FSHB
- Molecular Weight:37.95 kDa
- Sequence:NSCELTNITIAIEKEECRFCISINTTWCAGYCYTRDLVYKDPARPKIQKTCTFKELVYETVRVPGCAHHADSLYTYPVATQCHCGKCDSDSTDCTVRGLGPSYCSFGEMKE
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Tested Applications:Enzyme-linked immunoabsorbent assay, western blot (recombinant protein), antibody, protein array.
- Cat. No.:77539-544