Recombinant Human FGFR3 (from HEK293 cells)
Catalog # 77539-492
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:10 µg
- Storage Conditions:4 °C
- Endotoxin Content:<1 EU/μg by the LAL method
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:fibroblast growth factor receptor 3
- UniProtKB:P22607
- Protein/Peptide Name:FGFR3
- Purity:>95% as determined by Tris-Bis PAGE; >95% as determined by HPLC.
- Molecular Weight:The protein has a predicted MW of 41.4 kDa. Due to glycosylation, the protein migrates to 65 - 75 kDa based on bis-tris PAGE result.
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Cat. No.:77539-492