Recombinant Human CRYAB (from E. coli)
Vial, 500 µg
Catalog # 77539-178
Supplier: Creative Biomart
Supplier #: CRYAB-310H
Recombinant Human CRYAB (from <i>E. coli</i>)
Catalog # 77539-178
Supplier: Creative Biomart
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Specifications
Specifications
- Pack type:Vial
- Size:500 µg
- Storage Conditions:4 °C
- Protease-free:No
- Endotoxin Level:Low
- Animal-Free:No
- UniProtKB:P02511
- Purity:>95% by SDS-PAGE
- Carrier-Free:No
- Grade:Ultrapure grade
- Concentration:1 mg/ml (determined by Bradford assay)
- Source:E. coli
- Nuclease-free:No
- Cat. No.:77539-178
- Shipping Temperature:20
- Protein/Peptide Type:Recombinant
- Protein Synonyms:crystallin, alpha B
- Protein/Peptide Name:CRYAB
- Conjugation:Unconjugated
- Endotoxin-free:No
- Species:Human



