Recombinant Human BMP2 (from E. coli)
Catalog # 77538-750
Supplier: Creative Biomart
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:400 µg
- Storage Conditions:4 °C
- Endotoxin Content:<0.005 EU/μg protein (below level of detection)
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:bone morphogenetic protein 2
- UniProtKB:P12643
- Protein/Peptide Name:BMP2
- Molecular Weight:26 kDa (dimer)
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
- Tested Applications:iPSC and ESC maintenance and expansion, chemically defined media.
- Cat. No.:77538-750