Recombinant Human ATXN2 (from E. coli)
New Product
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:4 °C
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:ataxin 2
- UniProtKB:F8VQP2
- Protein/Peptide Name:ATXN2
- Purity:>80% by SDS-PAGE
- Molecular Weight:The protein has a calculated MW of 53.13 kDa.
- Sequence:MTPSGPVLASPQAGIIPTEAVAMPIPAASPTPASPASNRAVTPSSEAKDSRLQDQRQNSPAGNKENIKPNETSPSFSKAENKGISPVVSEHRKQIDDLKKFKNDFRLQPSSTSESMDQLLNKNREGEKSRDLIKDKIEPSAKDSFIENSSSNCTSGSSKPNSPSISPSILSNTEHKRGPEVTSQGVQTSSPACKQEKDDKEEKKDAAEQVRKSTLNPNAKEFNPRSFSQPKPSTTPTSPRPQAQPSPSMVGHQQPTPVYTQPVCFAPNMMYPVPVSPGVQPLYPIPMTPMPVNQAKTYRAVPNMPQQRQDQHHQSAMMHPASAAGPPIAATPPAYSTQYVAYSPQQFPNQPLVQHVPHYQSQHPHVYSPVIQGNARMMAPPTHAQPGLVSSSATQYGAHEQTHAMYACPKLPYNKETSPSFYFAISTGSLAQQYAHPNATLHPHTPHPQPSATPTGQQQSQHGGSHPAPSPVQHHQHQAAQLEHHHHHH
- Endotoxin Level:Low
- Concentration:0.5 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
Frequently Bought Together