Order Entry
United States
Orders LinkContactUsLinkComponent
 

Recombinant Human ASS1

Vial, 2 µg
Catalog # 77538-688
Supplier:   Creative Biomart
 :  ATG16L-945H
Recombinant Human ASS1
Catalog # 77538-688
Supplier:  Creative Biomart
CAS Number:  
Recombinant Human ASS1
Recombinant Human ASS1

Vial, 2 µg

CAS Number:  
Price
$530.89
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    2 µg
  • Storage Conditions:
    4 °C
  • Protease-free:
    No
  • Endotoxin Level:
    Low
  • Animal-Free:
    No
  • Carrier-Free:
    No
  • Grade:
    Ultrapure grade
  • Nuclease-free:
    No
  • Cat. No.:
    77538-688
  • Shipping Temperature:
    20
  • Protein/Peptide Type:
    Recombinant
  • Protein/Peptide Name:
    ASS1
  • Conjugation:
    Unconjugated
  • Endotoxin-free:
    No
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.