Recombinant Human ACTG2 (from E. coli)
Supplier: Creative Biomart
About this item
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 77538-548
- 77538-550
- Protein/Peptide Name:
- ACTG2
- ACTG2
- Protein Synonyms:
- actin, gamma 2, smooth muscle, enteric
- actin, gamma 2, smooth muscle, enteric
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Source:
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Sequence:
- MGSSHHHHHHSSGLVPRGSHMEEETTALVCDNGSGLCKAGFAGDDAPRAVFPSIVGRPRHQGVMVGMGQKDSYVGDEAQSKRGILTLKYPIEHGIITNWDDMEKIWHHSFYNELRVAPEEHPTLLTEAPLNPKANREKMTQIMFETFNVPAMYVAIQAVLSLYASGRTTGIVLDSGDGVTHNVPIYEGYALPHAIMRLDLAGRDLTDYLMKILTERGYSFVTTAEREIVRDIKEKLCYVALDFENEMATAASSSSLEKSYELPDGQVITIGNERFRCPETLFQPSFIGMESAGIHETTYNSIMKCDIDIRKDLYANNVLSGGTTMYPGIADRMQKEITALAPSTMKIKIIAPPERKYSVWIGGSILASLSTFQQMWISKPEYDEAGPSIVHRKCF
- MGSSHHHHHHSSGLVPRGSHMEEETTALVCDNGSGLCKAGFAGDDAPRAVFPSIVGRPRHQGVMVGMGQKDSYVGDEAQSKRGILTLKYPIEHGIITNWDDMEKIWHHSFYNELRVAPEEHPTLLTEAPLNPKANREKMTQIMFETFNVPAMYVAIQAVLSLYASGRTTGIVLDSGDGVTHNVPIYEGYALPHAIMRLDLAGRDLTDYLMKILTERGYSFVTTAEREIVRDIKEKLCYVALDFENEMATAASSSSLEKSYELPDGQVITIGNERFRCPETLFQPSFIGMESAGIHETTYNSIMKCDIDIRKDLYANNVLSGGTTMYPGIADRMQKEITALAPSTMKIKIIAPPERKYSVWIGGSILASLSTFQQMWISKPEYDEAGPSIVHRKCF
- Protease-free:
- No
- No
- Animal-Free:
- No
- No
- Nuclease-free:
- No
- No
- Carrier-Free:
- No
- No
- Endotoxin-free:
- No
- No
- Endotoxin Level:
- Low
- Low
- UniProtKB:
- P63267
- P63267
- Purity:
- >90%, by SDS-PAGE
- >90%, by SDS-PAGE
- Grade:
- Ultrapure grade
- Ultrapure grade
- Concentration:
- 0.2 mg/ml
- 0.2 mg/ml
- Molecular Weight:
- The protein has a calculated MW of 43.9 kDa.
- The protein has a calculated MW of 43.9 kDa.
- Storage Conditions:
- 4 °C
- 4 °C
- Shipping Temperature:
- 20
- 20
Specifications
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: ACTG2-3096H-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77538-548Protein/Peptide NameACTG2Protein Synonymsactin, gamma 2, smooth muscle, entericProtein/Peptide TypeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedSequenceMGSSHHHHHHSSGLVPRGSHMEEETTALVCDNGSGLCKAGFAGDDAPRAVFPSIVGRPRHQGVMVGMGQKDSYVGDEAQSKRGILTLKYPIEHGIITNWDDMEKIWHHSFYNELRVAPEEHPTLLTEAPLNPKANREKMTQIMFETFNVPAMYVAIQAVLSLYASGRTTGIVLDSGDGVTHNVPIYEGYALPHAIMRLDLAGRDLTDYLMKILTERGYSFVTTAEREIVRDIKEKLCYVALDFENEMATAASSSSLEKSYELPDGQVITIGNERFRCPETLFQPSFIGMESAGIHETTYNSIMKCDIDIRKDLYANNVLSGGTTMYPGIADRMQKEITALAPSTMKIKIIAPPERKYSVWIGGSILASLSTFQQMWISKPEYDEAGPSIVHRKCFProtease-freeNoAnimal-FreeNoNuclease-freeNoCarrier-FreeNoEndotoxin-freeNoEndotoxin LevelLowUniProtKBP63267Purity>90%, by SDS-PAGEGradeUltrapure gradeConcentration0.2 mg/mlMolecular WeightThe protein has a calculated MW of 43.9 kDa.Storage Conditions4 °CShipping Temperature20 - Supplier Number: ACTG2-3096H-200UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77538-550Protein/Peptide NameACTG2Protein Synonymsactin, gamma 2, smooth muscle, entericProtein/Peptide TypeRecombinantSpeciesHumanSourceE. coliConjugationUnconjugatedSequenceMGSSHHHHHHSSGLVPRGSHMEEETTALVCDNGSGLCKAGFAGDDAPRAVFPSIVGRPRHQGVMVGMGQKDSYVGDEAQSKRGILTLKYPIEHGIITNWDDMEKIWHHSFYNELRVAPEEHPTLLTEAPLNPKANREKMTQIMFETFNVPAMYVAIQAVLSLYASGRTTGIVLDSGDGVTHNVPIYEGYALPHAIMRLDLAGRDLTDYLMKILTERGYSFVTTAEREIVRDIKEKLCYVALDFENEMATAASSSSLEKSYELPDGQVITIGNERFRCPETLFQPSFIGMESAGIHETTYNSIMKCDIDIRKDLYANNVLSGGTTMYPGIADRMQKEITALAPSTMKIKIIAPPERKYSVWIGGSILASLSTFQQMWISKPEYDEAGPSIVHRKCFProtease-freeNoAnimal-FreeNoNuclease-freeNoCarrier-FreeNoEndotoxin-freeNoEndotoxin LevelLowUniProtKBP63267Purity>90%, by SDS-PAGEGradeUltrapure gradeConcentration0.2 mg/mlMolecular WeightThe protein has a calculated MW of 43.9 kDa.Storage Conditions4 °CShipping Temperature20



