Recombinant Human ACE2 (from HEK293 cells)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:4 °C
- Endotoxin Content:<1.0 EU/μg of the protein as determined by the LAL method
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized SARS-CoV-2 (2019-nCoV) Spike S1-His at 2 μg/ml (100 μl/well) can bind human ACE2 (hFc tag), the EC50 of ACE2 is 5 - 35 ng/ml.
- Reconstitution Instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-Free:No
- Protease-free:No
- Animal-Free:No
- Protein Synonyms:angiotensin I converting enzyme 2
- UniProtKB:Q9BYF1
- Protein/Peptide Name:ACE2
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The recombinant human ACE2/Fc is a disulfide-linked homodimeric protein. The reduced monomer consists of 961 amino acids and predicts a molecular mass of 110.3 kDa. As a result of glycosylation, the rhACE2/Fc monomer migrates as approximately 145 - 150 kDa band in SDS-PAGE under reducing conditions.
- Endotoxin Level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping Temperature:20
Specifications
Frequently Bought Together