Order Entry
United States
ContactUsLinkComponent
Acetyltransferase Assay Buffers
Acetyltransferase Assay Buffers
Supplier:  Sino Biological
New Product
Certificates
Acetyltransferase Assay Buffers
Supplier:  Sino Biological
New Product

Specifications

  • Storage Temperature:
    –20 °C
  • Formulation:
    Frozen solution of buffer containing: 250 mM Tris-HCl, pH 8.0, 0.5 mM EDTA and 25% glycerol. Add to final concentration of 2 mM DTT prior to use. DTT Solution supplied in separate vial.

Specifications

Related Information

Frequently Bought Together