Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
XFD532 NHS Ester
XFD532 NHS Ester [Same Structure to Alexa Fluor™ 532 NHS ester]
XFD532 NHS Ester [Same Structure to Alexa Fluor™ 532 NHS ester]
XFD532 NHS Ester [Same Structure to Alexa Fluor™ 532 NHS ester]
Catalog # 77463-082
Supplier:
AAT Bioquest
CAS Number:
Expand image
XFD532 NHS Ester [Same Structure to Alexa Fluor™ 532 NHS ester]
Catalog # 77463-082
Supplier:
AAT Bioquest
Supplier Number:
71904
CAS Number:
Specifications
Size:
5 mg
Cat. No.:
77463-082
Specifications
About this item
Product Details
:
:
723.77 g/mol
Product Details