$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
PE-Texas Red maleimide
PE-Texas Red maleimide, ReadiUse™, preactivated
PE-Texas Red maleimide, ReadiUse™, preactivated
PE-Texas Red maleimide, ReadiUse™, preactivated
Catalog # 77462-112
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
PE-Texas Red maleimide, ReadiUse™, preactivated
Catalog # 77462-112
Supplier:
AAT BIOQUEST INC
Supplier Number:
2716
CAS Number:
Specifications
Size:
1 mg
Cat. No.:
77462-112
Specifications
About this item
Product Details
:
Product Details