Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
APC-Cy5.5 maleimide
APC-Cy5.5 maleimide, ReadiUse™, preactivated
APC-Cy5.5 maleimide, ReadiUse™, preactivated
Catalog # 77462-122
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
APC-Cy5.5 maleimide, ReadiUse™, preactivated
Catalog # 77462-122
Supplier:
AAT BIOQUEST INC
Supplier Number:
2721
CAS Number:
Specifications
Size:
1 mg
Cat. No.:
77462-122
Specifications
About this item
Product Details
:
Product Details