Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
XFD532 SSL-NHS ester
XFD532 SSL-NHS ester, ReadiCleave™
XFD532 SSL-NHS ester, ReadiCleave™
XFD532 SSL-NHS ester, ReadiCleave™
Catalog # 77463-044
Supplier:
AAT Bioquest
CAS Number:
Expand image
XFD532 SSL-NHS ester, ReadiCleave™
Catalog # 77463-044
Supplier:
AAT Bioquest
Supplier Number:
7051
CAS Number:
Specifications
Size:
1 mg
Cat. No.:
77463-044
Specifications
About this item
Product Details
:
:
988.21 g/mol
Product Details