Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Psoralen TMP alkyne
Psoralen TMP alkyne
Psoralen TMP alkyne
Psoralen TMP alkyne
Catalog # 77462-252
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
Psoralen TMP alkyne
Catalog # 77462-252
Supplier:
AAT BIOQUEST INC
Supplier Number:
39063
CAS Number:
Specifications
Size:
1 mg
Cat. No.:
77462-252
Specifications
About this item
Product Details
:
Product Details