Protonex™ NIR 770 NHS ester
1 mg
Catalog # 77461-834
Supplier: AAT Bioquest
Supplier #: 21221
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Protonex™ NIR 770 NHS ester is the only cell-permeable acidotropic pH dye commercially available for developing NIR pH probes.
- Stable for 12 months
Quality tested in-house
Caution: For Research Use Only
- Molecular Weight:535.64 g/mol
Specifications
Specifications
- Size:1 mg